missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridinesulfonic acid, 97%, Thermo Scientific Chemicals

Catalog No. AC455760050
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455760050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. AC455760050 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 455760050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15103-48-7
Molecular Formula C5H5NO3S
Molecular Weight (g/mol) 159.16
MDL Number MFCD00234080
InChI Key KZVLNAGYSAKYMG-UHFFFAOYSA-N
Synonym 2-pyridinesulfonic acid, pyridinesulfonic acid, 2-pyridinesulfonic acid 8ci 9ci, alpha-pyridinesulfonic acid, sulfopyridin, pyridine-2-sulfonicacid, acmc-1bo1h, .alpha.-pyridinesulfonic acid, kzvlnagysakymg-uhfffaoysa
PubChem CID 94849
IUPAC Name pyridine-2-sulfonic acid
SMILES OS(=O)(=O)C1=CC=CC=N1

Specifications

Melting Point 244.0°C to 249.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: very soluble.
Formula Weight 159.16
Percent Purity 97%
Chemical Name or Material 2-Pyridinesulfonic acid
missing translation for 'healthHazard1' GHS Signal Word: Danger
missing translation for 'healthHazard2' GHS H Statement
Causes severe skin burns and eye damage.
missing translation for 'healthHazard3' GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately

RUO – Research Use Only