Learn More
2-Pyridin-3-ylpiperidine, 97%
CAS: 13078-04-1 | C10H14N2 | 162.24 g/mol
$520.23 - $520.23
Chemical Identifiers
| CAS | 13078-04-1 |
|---|---|
| Molecular Formula | C10H14N2 |
| Molecular Weight (g/mol) | 162.24 |
| MDL Number | MFCD00006370 |
| InChI Key | MTXSIJUGVMTTMU-UHFFFAOYNA-N |
| Synonym | 3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl |
| PubChem CID | 2181 |
| ChEBI | CHEBI:28986 |
| IUPAC Name | 3-piperidin-2-ylpyridine |
| SMILES | C1CCC(NC1)C1=CC=CN=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC458980010
|
Thermo Scientific Chemicals
458980010 |
1 g |
Each for $520.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13078-04-1 | |
| 162.24 | |
| MTXSIJUGVMTTMU-UHFFFAOYNA-N | |
| 2181 | |
| 3-piperidin-2-ylpyridine |
| C10H14N2 | |
| MFCD00006370 | |
| 3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl | |
| CHEBI:28986 | |
| C1CCC(NC1)C1=CC=CN=C1 |
Specifications
| 13078-04-1 | |
| 1.0460g/mL | |
| 93°C | |
| 96% min. (GC) | |
| C10H14N2 | |
| 1 g | |
| 3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl | |
| C1CCC(NC1)C1=CC=CN=C1 | |
| 162.24 | |
| CHEBI:28986 | |
| 97% |
| 9.0°C | |
| 270.0°C to 272.0°C | |
| Authentic | |
| Glass Bottle | |
| MFCD00006370 | |
| 1.046 | |
| MTXSIJUGVMTTMU-UHFFFAOYNA-N | |
| 3-piperidin-2-ylpyridine | |
| 2181 | |
| 162.23 | |
| 2-Pyridin-3-ylpiperidine |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
Danger
RUO – Research Use Only