accessibility menu, dialog, popup

UserName

NHN

CAS RN 13078-04-1

IUPAC Name: 3-(piperidin-2-yl)pyridine
Synonyms: anabasine (+-)-anabasine 八角枫碱 DL-anabasine (+/-)-anabasine
Molecular Weight (g/mol): 162.24
Molecular Formula: C10H14N2
InChi Key: MTXSIJUGVMTTMU-UHFFFAOYNA-N
SMILES: C1CCC(NC1)C1=CC=CN=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results