missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridin-3-ylpiperidine, 97%

Catalog No. AC458980050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458980050 5 g
AC458980010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458980050 Supplier Thermo Scientific Chemicals Supplier No. 458980050

CAS: 13078-04-1 | C10H14N2 | 162.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13078-04-1
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.24
MDL Number MFCD00006370
InChI Key MTXSIJUGVMTTMU-UHFFFAOYNA-N
Synonym 3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl
PubChem CID 2181
ChEBI CHEBI:28986
IUPAC Name 3-piperidin-2-ylpyridine
SMILES C1CCC(NC1)C1=CC=CN=C1

Specifications

Melting Point 9.0°C
Density 1.0460g/mL
Boiling Point 270.0°C to 272.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.046
Formula Weight 162.23
Percent Purity 97%
Chemical Name or Material 2-Pyridin-3-ylpiperidine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre

RUO – Research Use Only