missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrid-4-ylpyrimidine-5-carboxylic acid, 90%, Thermo Scientific™

Catalog No. p-3586683
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC66601CB 250mg Amber Glass Bottle
CC66601DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC66601CB Supplier Thermo Scientific Chemicals Supplier No. CC66601CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 259807-47-1
Molecular Formula C10H7N3O2
Molecular Weight (g/mol) 201.185
MDL Number MFCD09863218
InChI Key GTKJGOOMSODGOU-UHFFFAOYSA-N
Synonym 2-pyridin-4-yl pyrimidine-5-carboxylic acid, 2-pyrid-4-ylpyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-4-pyridinyl, 2-4-pyridyl pyrimidine-5-carboxylic acid, 2-4-pyridinyl-5-pyrimidinecarboxylic acid, 2-pyridin-4-yl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylic acid, 2-4-pyridinyl, 5-pyrimidinecarboxylic acid, 2-4-pyridinyl-9ci
PubChem CID 22710213
IUPAC Name 2-pyridin-4-ylpyrimidine-5-carboxylic acid
SMILES C1=CN=CC=C1C2=NC=C(C=N2)C(=O)O

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 201.18
Percent Purity 90%
Chemical Name or Material 2-Pyrid-4-ylpyrimidine-5-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.