missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrazinecarboxylic acid, 99%

Catalog No. p-188787
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131710250 25 g Plastic bottle
AC131711000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC131710250 Supplier Thermo Scientific Chemicals Supplier No. 131710250
Only null left

CAS: 98-97-5 | C5H4N2O2 | 124.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-97-5
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.1
MDL Number MFCD00006130
InChI Key NIPZZXUFJPQHNH-UHFFFAOYSA-N
Synonym 2-pyrazinecarboxylic acid, pyrazinoic acid, pyrazinecarboxylic acid, pyrazinic acid, 2-carboxypyrazine, paradiazinecarboxylic acid, pyrazinemonocarboxylic acid, 2-pyrazinoic acid, 2-pyrazine carboxylic acid, piazinecarboxylic acid
PubChem CID 1047
ChEBI CHEBI:71311
IUPAC Name pyrazine-2-carboxylic acid
SMILES C1=CN=C(C=N1)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 222.0°C to 225.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 25 g
Beilstein 25, 125
Merck Index 15, 8070
Solubility Information Solubility in water: soluble in cold water. Other solubilities: 1g/120g absolubte ethanol (25 c),practically insoluble in ether,chloroform and,benzene
Formula Weight 124.1
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Pyrazinecarboxylic acid, 99%
Show More Show Less
EINECSNumber 202-718-1
TSCA TSCA

RUO – Research Use Only