missing translation for 'onlineSavingsMsg'
Learn More

2-Propynyl [3-(Triethoxysilyl)propyl]carbamate 90.0+%, TCI America™

Catalog No. P22585G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22585G 5 g
P22581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22585G Supplier TCI America Supplier No. P22585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 870987-68-1
Molecular Formula C13H25NO5Si
Molecular Weight (g/mol) 303.43
MDL Number MFCD01320971
InChI Key ZMASSMHNNATIBZ-UHFFFAOYSA-N
Synonym [3-(Triethoxysilyl)propyl]carbamic Acid 2-Propynyl Ester, Propargyl [3-(Triethoxysilyl)propyl]carbamate, [3-(Triethoxysilyl)propyl]carbamic Acid Propargyl Ester
PubChem CID 59324374
IUPAC Name prop-2-ynyl N-(3-triethoxysilylpropyl)carbamate
SMILES CCO[Si](CCCNC(=O)OCC#C)(OCC)OCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 303.43
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propynyl [3-(Triethoxysilyl)propyl]carbamate