missing translation for 'onlineSavingsMsg'
Learn More

2-Propyn-1-ol 98.0+%, TCI America™

Catalog No. p-7134665
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P053625ML 25 mL
P0536100ML 100 mL
P0536500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P053625ML Supplier TCI America Supplier No. P053625ML
Only null left

Chemical Identifiers

CAS 107-19-7
Molecular Formula C3H4O
Molecular Weight (g/mol) 56.06
MDL Number MFCD00002912
InChI Key TVDSBUOJIPERQY-UHFFFAOYSA-N
Synonym propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne
PubChem CID 7859
ChEBI CHEBI:28905
IUPAC Name prop-2-yn-1-ol
SMILES OCC#C

Specifications

Melting Point -52°C
Color Yellow
pH 7 (50g/L H2O soln. 20°C)
Boiling Point 113°C
Quantity 25 mL
UN Number 2929
Formula Weight 56.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propyn-1-ol
EINECSNumber (2)-0272
RTECSNumber UK5075000
TSCA Yes
Recommended Storage Refrigerator