missing translation for 'onlineSavingsMsg'
Learn More

2-Propylvaleric Acid 99.0+%, TCI America™

Supplier:  TCI America P0823500ML

 View more versions of this product

Catalog No. P0823500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Propylvaleric Acid
Yellow
C8H16O2
500 mL
NIJJYAXOARWZEE-UHFFFAOYSA-N
2-propylpentanoic acid
3121
144.21
Liquid
99-66-1
220°C
MFCD00002672
valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl
CCCC(CCC)C(O)=O
144.21
CHEBI:39867
≥99.0% (GC,T)

Chemical Identifiers

99-66-1
144.21
NIJJYAXOARWZEE-UHFFFAOYSA-N
3121
2-propylpentanoic acid
C8H16O2
MFCD00002672
valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl
CHEBI:39867
CCCC(CCC)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : YV7875000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions