Learn More
2-Propylpentanoic acid, sodium salt, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 271830250
| Quantity | 25g |
|---|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C8H15NaO2 | |
| MFCD00078604 | |
| sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin | |
| CHEBI:9925 | |
| CCCC(CCC)C(=O)[O-].[Na+] |
Specifications
| 2-Propylpentanoic acid, sodium salt | |
| 1069-66-5 | |
| 100.0 | |
| 98% | |
| C8H15NaO2 | |
| 25g | |
| 15, 10099 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol | |
| CCCC(CCC)C(=O)[O-].[Na+] | |
| 166.2 | |
| CHEBI:9925 | |
| 98% |
| 98% | |
| 97.5 | |
| Authentic | |
| Glass bottle | |
| (CH3CH2CH2)2CHCOONa | |
| MFCD00078604 | |
| sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin | |
| AEQFSUDEHCCHBT-UHFFFAOYSA-M | |
| sodium;2-propylpentanoate | |
| 16760703 | |
| 166.2 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May damage the unborn child.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Danger
EINECSNumber : 213-961-8
RTECSNumber : YV7876000
TSCA : TSCA