missing translation for 'onlineSavingsMsg'
Learn More

2-Propylfuran 98.0+%, TCI America™

Catalog No. p-7135156
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14885G Supplier TCI America Supplier No. P14885G

Chemical Identifiers

CAS 4229-91-8
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
MDL Number MFCD00047070
InChI Key CPLJMYOQYRCCBY-UHFFFAOYSA-N
Synonym furan, 2-propyl, 2-n-propylfuran, 2-n-propyl furan, furan, propyl, 2-propyl-furan, furan, alpha-propyl, acmc-1ctud, 2-propyl furan, furan, .alpha.-propyl, cpljmyoqyrccby-uhfffaoysa
PubChem CID 77907
IUPAC Name 2-propylfuran
SMILES CCCC1=CC=CO1

Specifications

Color Yellow
Boiling Point 115°C
Quantity 5 g
UN Number 1993
Formula Weight 110.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Propylfuran