missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propylaniline 98.0+%, TCI America™
$437.02 - $1678.75
Chemical Identifiers
| CAS | 1821-39-2 |
|---|---|
| Molecular Formula | C9H13N |
| Molecular Weight (g/mol) | 135.21 |
| MDL Number | MFCD00009803 |
| InChI Key | WKURVXXDGMYSDP-UHFFFAOYSA-N |
| PubChem CID | 15759 |
| IUPAC Name | 2-propylaniline |
| SMILES | CCCC1=CC=CC=C1N |
Chemical Identifiers
| 1821-39-2 | |
| 135.21 | |
| WKURVXXDGMYSDP-UHFFFAOYSA-N | |
| 2-propylaniline |
| C9H13N | |
| MFCD00009803 | |
| 15759 | |
| CCCC1=CC=CC=C1N |
Specifications
| 1821-39-2 | |
| 223°C | |
| MFCD00009803 | |
| WKURVXXDGMYSDP-UHFFFAOYSA-N | |
| 2-propylaniline | |
| 15759 | |
| ≥98.0% (GC,T) | |
| 2-Propylaniline |
| Yellow | |
| C9H13N | |
| 5 g | |
| CCCC1=CC=CC=C1N | |
| 135.21 | |
| 135.21 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0206
RTECSNumber : BY8300000
TSCA : No