missing translation for 'onlineSavingsMsg'
Learn More

2-Propoxyethanol, 98%, pure

Catalog No. p-3758722
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass Bottle
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418400010 1 kg Glass Bottle
AC418400050 5 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC418400010 Supplier Thermo Scientific Chemicals Supplier No. 418400010

CAS: 2807-30-9 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2807-30-9
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00072645
InChI Key YEYKMVJDLWJFOA-UHFFFAOYSA-N
Synonym ethanol, 2-propoxy, ethylene glycol monopropyl ether, propyl cellosolve, propoxyethanol, propyl glycol, 2-propoxyethan-1-ol, ektasolve ep, 2-propyloxy ethanol, n-propoxyethanol, ethylene glycol mono-n-propyl ether
PubChem CID 17756
IUPAC Name 2-propoxyethanol
SMILES CCCOCCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 149.0°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.4120 to 1.4140
Linear Formula CH3(CH2)2OCH2CH2OH
Quantity 1 kg
Specific Gravity 0.91
Solubility Information Solubility in water: soluble.
Formula Weight 104.15
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2-Propoxyethanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Toxic in contact with skin.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Keep away from heat/spark
EINECSNumber 220-548-6

RUO – Research Use Only