missing translation for 'onlineSavingsMsg'
Learn More

2-Propoxybenzoic Acid 98.0+%, TCI America™

Catalog No. p-7135213
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16945G 5 g
P169425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16945G Supplier TCI America Supplier No. P16945G

Chemical Identifiers

CAS 2100-31-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD01075687
InChI Key OXOWWPXTTOCKKU-UHFFFAOYSA-N
Synonym 2-n-propoxybenzoic acid, o-propoxybenzoic acid, benzoic acid, 2-propoxy, 2-propoxy benzoic acid, salicylic acid propyl ether, akos bbb/270, rarechem al bo 0876, propyloxybenzoic acid, 2-propoxybenzoic acid, o-propoxy benzoic acid
PubChem CID 539215
IUPAC Name 2-propoxybenzoic acid
SMILES CCCOC1=CC=CC=C1C(=O)O

Specifications

Melting Point 39°C
Boiling Point 207°C
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Propoxybenzoic Acid
TSCA No
Recommended Storage Refrigerator