missing translation for 'onlineSavingsMsg'
Learn More

2-Propionylthiazole, 99%

CAS: 43039-98-1 | C6H7NOS | 141.188 g/mol

Supplier:  Thermo Scientific Chemicals B2171109

 View more versions of this product

Catalog No. AAB2171109


Only null left
Explore available promotions

Description

Description

Used as flavoring agent and fragrance chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as flavoring agent and fragrance chemical compounds.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

2-Propionylthiazole
1.174
>100°C (212°F)
MFCD01681404
2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k
TYRAENAWSLPSLW-UHFFFAOYSA-N
1-(1,3-thiazol-2-yl)propan-1-one
65288
99%
43039-98-1
110°C (5 mmHg)
C6H7NOS
10 g
Not miscible or difficult to mix with water.
CCC(=O)C1=NC=CS1
141.188
141.19

Chemical Identifiers

43039-98-1
141.188
TYRAENAWSLPSLW-UHFFFAOYSA-N
65288
CCC(=O)C1=NC=CS1
C6H7NOS
MFCD01681404
2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k
1-(1,3-thiazol-2-yl)propan-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : XJ5123000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only