Learn More
2-Propionylthiazole, 99%
CAS: 43039-98-1 | C6H7NOS | 141.188 g/mol
Supplier: Thermo Scientific Chemicals B2171104
Description
Used as flavoring agent and fragrance chemical compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as flavoring agent and fragrance chemical compounds.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 2-Propionylthiazole | |
| 1.174 | |
| >100°C (212°F) | |
| MFCD01681404 | |
| 2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k | |
| TYRAENAWSLPSLW-UHFFFAOYSA-N | |
| 1-(1,3-thiazol-2-yl)propan-1-one | |
| 65288 | |
| 99% |
| 43039-98-1 | |
| 110°C (5 mmHg) | |
| C6H7NOS | |
| 2 g | |
| Not miscible or difficult to mix with water. | |
| CCC(=O)C1=NC=CS1 | |
| 141.188 | |
| 141.19 |
Chemical Identifiers
| 43039-98-1 | |
| 141.188 | |
| TYRAENAWSLPSLW-UHFFFAOYSA-N | |
| 65288 | |
| CCC(=O)C1=NC=CS1 |
| C6H7NOS | |
| MFCD01681404 | |
| 2-propionylthiazole, 1-thiazol-2-yl propan-1-one, 2-propionyl thiazole, thiazole, 2-propionyl, 1-propanone, 1-2-thiazolyl, 1-2-thiazolyl-1-propanone, ethyl 2-thiazolyl ketone, 1-1,3-thiazol-2-yl propan-1-one, 2-propionylthiazol, unii-05cb62uh9k | |
| 1-(1,3-thiazol-2-yl)propan-1-one |
Safety and Handling
RTECSNumber : XJ5123000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only