missing translation for 'onlineSavingsMsg'
Learn More

2-Propionylfuran, 97%

Catalog No. AAH2675414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2675414 25 g
AAH2675406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2675414 Supplier Thermo Scientific Chemicals Supplier No. H2675414
Only null left

CAS: 3194-15-8 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3194-15-8
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00020936
InChI Key HCPORNAVHSWTOJ-UHFFFAOYSA-N
Synonym 2-propionylfuran, 1-2-furyl propan-1-one, 1-furan-2-yl propan-1-one, 1-propanone, 1-2-furanyl, 2-furyl ethyl ketone, ethyl 2-furyl ketone, furyl ethyl ketone, 1-propanone, 1-2-furyl, ethyl furyl ketone, 2-propanoylfuran
PubChem CID 76662
IUPAC Name 1-(furan-2-yl)propan-1-one
SMILES CCC(=O)C1=CC=CO1

Specifications

Melting Point 26°C to 30°C
Boiling Point 53°C to 55°C (1 mmHg)
Odor Characteristic
Quantity 25 g
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 2-Propionylfuran
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 221-693-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only