CAS RN 3194-15-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Propionylfuran, 97%
CAS: 3194-15-8 Molecular Formula: C7H8O2 Molecular Weight (g/mol): 124.14 MDL Number: MFCD00020936 InChI Key: HCPORNAVHSWTOJ-UHFFFAOYSA-N Synonym: 2-propionylfuran,1-2-furyl propan-1-one,1-furan-2-yl propan-1-one,1-propanone, 1-2-furanyl,2-furyl ethyl ketone,ethyl 2-furyl ketone,furyl ethyl ketone,1-propanone, 1-2-furyl,ethyl furyl ketone,2-propanoylfuran PubChem CID: 76662 IUPAC Name: 1-(furan-2-yl)propan-1-one SMILES: CCC(=O)C1=CC=CO1