missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Propargyloxy)benzaldehyde 98.0+%, TCI America™
Supplier: TCI America P23381G
Specifications
| 2-(Propargyloxy)benzaldehyde | |
| 70°C | |
| MFCD02380378 | |
| 2-(2-Propynyloxy)benzaldehyde | |
| C#CCOC1=CC=CC=C1C=O | |
| 160.172 | |
| 160.17 | |
| Crystalline Powder |
| 29978-83-4 | |
| C10H8O2 | |
| 1 g | |
| PEERMPFPCWSJLC-UHFFFAOYSA-N | |
| 2-prop-2-ynoxybenzaldehyde | |
| 583037 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 29978-83-4 | |
| 160.172 | |
| PEERMPFPCWSJLC-UHFFFAOYSA-N | |
| 583037 | |
| C#CCOC1=CC=CC=C1C=O |
| C10H8O2 | |
| MFCD02380378 | |
| 2-(2-Propynyloxy)benzaldehyde | |
| 2-prop-2-ynoxybenzaldehyde |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator