missing translation for 'onlineSavingsMsg'
Learn More

2-(Propargyloxy)benzaldehyde 98.0+%, TCI America™

Catalog No. p-7135525
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2338200MG 200 mg
P23381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2338200MG Supplier TCI America Supplier No. P2338200MG

Chemical Identifiers

CAS 29978-83-4
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD02380378
InChI Key PEERMPFPCWSJLC-UHFFFAOYSA-N
Synonym 2-(2-Propynyloxy)benzaldehyde
PubChem CID 583037
IUPAC Name 2-prop-2-ynoxybenzaldehyde
SMILES C#CCOC1=CC=CC=C1C=O

Specifications

Melting Point 70°C
Quantity 200 mg
Formula Weight 160.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Propargyloxy)benzaldehyde
TSCA No
Recommended Storage Refrigerator