missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, Semiconductor Grade, 99.5% min

Maximum level of impurities: Color (APHA) 10, Evaporation residue 5ppm, Solubility in H2O P.T., H2O (by Karl Fischer titrn) 0.05%, Acidity 0.2meq/g, Alkalinity 0.1meq/g, Cl 0.2ppm, PO4 0.5ppm | CAS: 67-63-0 | C3H8O | 60.096 g/mol

$85.97 - $970.92

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA19397AP
View Documents
Thermo Scientific Chemicals
019397AP
500 mL
Each for $85.97
Only null left
 
AA19397K2
View Documents
Thermo Scientific Chemicals
019397K2
1 L
Each for $118.99
Only null left
 
AA19397M1
View Documents
Thermo Scientific Chemicals
019397M1
4 x 1 L
Each for $347.77
Only null left
 
AA19397K7
View Documents
Thermo Scientific Chemicals
019397K7
4 L
Each for $285.99
Only null left
 
AA19397M6
View Documents
Thermo Scientific Chemicals
019397M6
4 x 4 L
Each for $970.92
Only null left
 
Description

Description

2-Propanol is used as a solvent for gums, resins, alkaloids and essential oils, as a denaturant for ethyl alcohol and is used in lotions and cosmetics. It is used in keyboard, LCD and laptop cleaning, as a wetting agent in the fountain solution used in lithographic printing, and often used as a solvent for French polishing shellac used in cabinet making. It acts as hydride source in the Meerwein-Ponndorf-Verley reduction. It plays an important role in DNA extraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
MFCD00011674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O

Specifications

67-63-0
Colorless
82°C to 83°C
455°C (851°F)
Alcohol-like
1.377
500 mL
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1
Semiconductor
Liquid
°C to 90°C
0.785
12°C (53°F)
99.5% min.
C3H8O
MFCD00011674
635639
Miscible with water,ethanol,acetone,chloroform,methanol,diethyl ether,n-octanol and benzene.
CC(C)O
60.096
CHEBI:17824
48 hPa (36mm Hg at 20°C)
2-Propanol
2-Propanol
Videos
Safety and Handling

Safety and Handling

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c

H225-H319-H335-H336-H373

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 200-661-7

RTECSNumber : NT8050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only