missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, CHROMASOLV™ LC-MS, Solstice

Catalog No. 60046511
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046511 4 x 2.5 L
6000134 1 L
6000135 2.5 L
60046512 6 x 1 L
6000136 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60046511 Supplier Solstice Supplier No. 349654X2.5L

CHROMASOLV™ LC-MS

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -89.5°C (literature)
CAS Max % 100.0000%
Density 0.785 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Flash Point 12°C
Packaging Glass Bottle
Linear Formula (CH3)2CHOH
UN Number UN1219
Beilstein 635639
Boiling Point 77°C to 83°C
Formula Weight 60.11
Percent Purity >99.9%
Grade LC-MS
Chemical Name or Material 2-Propanol, CHROMASOLV™ LC-MS
Show More Show Less
ShelfLife 360 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol
EINECSNumber 200-661-7
Recommended Storage Room Temp