missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, CHROMASOLV™ LC-MS, Solstice

Catalog No. 6000136
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000136 4 x 4 L
6000134 1 L
6000135 2.5 L
60046512 6 x 1 L
60046511 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6000136 Supplier Solstice Supplier No. 349654X4L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™ LC-MS

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Density 0.785g/mL at 25°C
Quantity 4 x 4 L
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Linear Formula (CH3)2CHOH
Vapor Pressure 44 mmHg (25°C), 33 mmHg (20°C)
UN Number UN1219
Beilstein 635639
Formula Weight 60.1g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
RTECSNumber NT8050000