missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol (HPLC), Fisher Chemical™

Catalog No. p-216301
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Safe-Cote™ Amber Glass 
Wide Neck Amber Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A4511 1 L Wide Neck Amber Bottle
A4514 4 L Glass Bottle
A451SK4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A4511 Supplier Thermo Fisher Scientific Supplier No. A4511
Only null left
Edge

HPLC mobile phase for analytical and preparative separations

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH 7
Melting Point -89.5°C
Absorbance 0.025 AU max. at 254nm, 0.13 AU max. at 230nm, 0.25 AU max. at 220nm, 1.00 AU max. at 205nm
Color Colorless
Density 0.785g/cm³
Quantity 1 L
Identification Pass Test
Packaging Wide Neck Amber Bottle
Physical Form Liquid
Refractive Index 1.3740 to 1.3760 (at 25°C)
Assay Percent Range 99.8% min by GC
Residue after Evaporation 2ppm max.
Vapor Pressure 43mmHg at 20°C
Viscosity 2.27 mPa/s at 20°C
Water 0.10% max.
Boiling Point 81°C
Formula Weight 60.1
Percent Purity ≥99.9%
Grade Mobile phase for HPLC applications
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid

Safe-Cote™ bottles that provide the purity of glass and most of the benefits for storing and dispensing chemicals. If they break, glass fragments and liquids are more likely to remain trapped.