missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, Honeywell™
Semiconductor Grade VLSI (Honeywell 17604)
$1085.18 - $1085.18
Chemical Identifiers
| CAS | 67-63-0 |
|---|---|
| Molecular Formula | C3H8O |
| Molecular Weight (g/mol) | 60.10 |
| InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
| PubChem CID | 3776 |
| ChEBI | CHEBI:17824 |
| IUPAC Name | propan-2-ol |
| SMILES | CC(C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
60-090-335
|
Honeywell Inc
40301H25L |
2.5 L |
Each for $1,085.18
|
|
|||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Safety and Handling
Recommended Storage : Room Temperature