missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, GC Grade, 99.7+%

Catalog No. p-4408284
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40983K2 1 L
AA40983M1 4 x 1 L
AA40983K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40983K2 Supplier Thermo Scientific Chemicals Supplier No. 040983K2
Only null left

Suitable For Environmental, Pesticide, and GC Analysis | CAS: 67-63-0 | C3H8O | 60.096 g/mol

Suitable for environmental and GC analysis. Used in making cosmetics, skin and hair preparations, pharmaceuticals, perfumes, lacquer formulations, dye solutions, anti freezes, soaps and window cleaners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point °C to 90°C
Color Colorless
Appearance Colorless liquid
Density 0.785 g/cm3 at 20°C
Quantity 1 L
Flash Point 12°C (53°F)
Ignition Point 455°C (851°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.377
Assay Percent Range >99.7%
Vapor Pressure 48 hPa (36mm Hg) at 20°C
Beilstein 635639
Boiling Point 82°C to 83°C
Solubility Information Miscible in water,benzene,chloroform,ethanol,ether,glycerine and acetone.
Formula Weight 60.1
Grade GC
Chemical Name or Material 2-Propanol, GC Grade
Show More Show Less
Hazard Category H225-H319-H335-H336-H373
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 200-661-7
RTECSNumber NT8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only