missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, For pesticide residue analysis, Honeywell Riedel-de Haën™

Catalog No. p-7072101
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
4 x 4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000138 2.5 L
60046347 4 x 2.5 L
6000142 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000138 Supplier Honeywell Inc Supplier No. 344862.5L
Only null left

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Density 0.785g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Linear Formula (CH3)2CHOH
Vapor Pressure 33 mmHg (20°C), 44 mmHg (25°C)
UN Number UN1219
Beilstein 635639
Formula Weight 60.1g/mol
Percent Purity ≥99.8% (GC)
Grade CHROMASOLV
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
RTECSNumber NT8050000