missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™

Catalog No. M1009981000
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1009981000 1 L Glass Bottle
M1009982500 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1009981000 Supplier Technidata Supplier No. 1.00998.1000

For gas chromatography ECD and FID SupraSolv™

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH Neutral (at 20°C)
Melting Point -89.5°C
Color Colorless
Density 0.78g/cm3 at 20°C
Quantity 1 L
Flash Point 12°C (54°F)
Ignition Point 425°C (797.9°F)
Odor Alcohol-like
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.37
Vapor Pressure 43 hPa (20°C)
UN Number 1219
Boiling Point 82.4°C
Solubility Information Soluble in water at 20°C
Formula Weight 60.1
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H319: Causes serious eye irritation.
Precautionary Statement H336: May cause drowsiness or dizziness.
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.