missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, CHROMASOLV™ Plus, for HPLC, 99.9%, Honeywell Riedel-de Haën™
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Specifications
Specifications
| Density | 0.785g/mL at 25°C |
| Quantity | 1 L |
| Explosion Limit | 0.02 |
| Flash Point | 12°C (53.6°F) |
| Ignition Point | 750°F |
| Linear Formula | (CH3)2CHOH |
| Vapor Pressure | 33 mmHg (20°C), 44 mmHg (25°C) |
| UN Number | UN1219 |
| Beilstein | 635639 |
| Formula Weight | 60.1g/mol |
| Show More |
Safety and Handling
| Hazard Statement | P210-P261-P305 + P351 + P338 |
| RTECSNumber | NT8050000 |