missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, CHROMASOLV™ Plus, for HPLC, 99.9%, Solstice

Catalog No. 6000131
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000131 1 L
6000132 2.5 L
6000133 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000131 Supplier Solstice Supplier No. 6504471L

CHROMASOLV™ Plus, for HPLC, 99.9%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Density 0.785g/mL at 25°C
Quantity 1 L
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Vapor Pressure 33 mmHg (20°C), 44 mmHg (25°C)
Linear Formula (CH3)2CHOH
UN Number UN1219
Beilstein 635639
Formula Weight 60.1g/mol
Percent Purity 99%
Grade CHROMASOLV™ Plus
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
RTECSNumber NT8050000