missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol CHROMASOLV™, for HPLC, 99.9%, Honeywell Riedel-de Haën™

Catalog No. p-7073735
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
185 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000123 1 L
6000124 2.5 L
6000130 6 x 1 L
6000128 4 x 2.5 L
6000122 185 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6000123 Supplier Honeywell Chemicals Supplier No. 348631L
Only null left

CHROMASOLV™, for HPLC, 99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -89.5°C
Density 0.785g/mL at 25°C
Boiling Point 82°C
Quantity 1 L
Refractive Index n20/D 1.377
Linear Formula (CH3)2CHOH
Vapor Pressure 44mmHg (25°C)
Beilstein 635639
Formula Weight 60.1g/mol
Percent Purity 99.9%
Grade CHROMASOLV
Chemical Name or Material 2-Propanol, for HPLC
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
Precautionary Statement H225-H319-H336
EINECSNumber 200-661-7
RTECSNumber NT8050000