missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol CHROMASOLV™, for HPLC, 99.9%, Solstice
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Melting Point | -89.5°C |
| Density | 0.785g/mL at 25°C |
| Quantity | 2.5 L |
| Refractive Index | n20/D 1.377 |
| Linear Formula | (CH3)2CHOH |
| Vapor Pressure | 44mmHg (25°C) |
| Beilstein | 635639 |
| Boiling Point | 82°C |
| Formula Weight | 60.1g/mol |
| Percent Purity | 99.9% |
| Show More |
Safety and Handling
| Hazard Statement | P210-P261-P305 + P351 + P338 |
| Precautionary Statement | H225-H319-H336 |
| EINECSNumber | 200-661-7 |
| RTECSNumber | NT8050000 |