missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, CHROMASOLV™ LC-MS, Solstice
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Specifications
Specifications
| Color | Colorless |
| Density | 0.785g/mL at 25°C |
| Quantity | 2.5 L |
| Explosion Limit | 0.02 |
| Flash Point | 12°C (53.6°F) |
| Ignition Point | 750°F |
| Linear Formula | (CH3)2CHOH |
| Vapor Pressure | 44 mmHg (25°C), 33 mmHg (20°C) |
| UN Number | UN1219 |
| Beilstein | 635639 |
| Show More |
Safety and Handling
| Hazard Statement | P210-P261-P305 + P351 + P338 |
| RTECSNumber | NT8050000 |