missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020417
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020417 1 mL
111018964 5 mL
111018533 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111020417 Supplier Merck Emd Millipore Supplier No. 912371MLF

2-Propanol is a secondary alcohol, which can find a wide number of applications as a solvent.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 67-63-0
Ignition Point 750°F
Vapor Pressure 44 mmHg (25°C)
Vapor Density 2.1 (vs. air)
Density 0.785 g/mL (at 25°C)
Flash Point 12°C
Physical Form Neat
Linear Formula (CH3)2 CHOH
MDL Number MFCD00011674
Melting Point -89.5°C
Boiling Point 82°C
Synonym sec-Propyl alcohol; IPA; Isopropanol; Isopropyl alcohol
Solubility Completely soluble in water
Type Analytical Standard
Quantity 1 mL
Formula Weight 60.10
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less