missing translation for 'onlineSavingsMsg'
Learn More

2-Propanethiol, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019642
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019642 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019642 Supplier Merck Emd Millipore Supplier No. 407211ML
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 75-33-2
Vapor Pressure 455 mmHg (37.8°C)
Vapor Density 2.6 (vs. air)
Density 0.82 g/mL (at 25°C)
Linear Formula (CH3)2 CHSH
MDL Number MFCD00004863
Melting Point -131°C
Boiling Point 57°C to 60°C
Synonym Isopropyl mercaptan
Type Analytical Standard
Quantity 1 mL
Formula Weight 76.16
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Show More Show Less