missing translation for 'onlineSavingsMsg'
Learn More

2-Propanesulfonyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 222030250

 View more versions of this product

Catalog No. AC222030250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Propanesulfonyl chloride
1.2730g/mL
86°C
Glass bottle
(CH3)2CHSO2Cl
04,8
isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride
CC(C)S(=O)(=O)Cl
142.61
142.61
10147-37-2
95.0°C (45.0 mmHg)
96% min. (GC)
C3H7ClO2S
25g
1.273
DRINJBFRTLBHNF-UHFFFAOYSA-N
propane-2-sulfonyl chloride
82408
97%

Chemical Identifiers

10147-37-2
142.61
isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride
propane-2-sulfonyl chloride
C3H7ClO2S
DRINJBFRTLBHNF-UHFFFAOYSA-N
82408
CC(C)S(=O)(=O)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 233-415-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only