missing translation for 'onlineSavingsMsg'
Learn More

2-Propanesulfonyl chloride, 97%

Catalog No. AC222030050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222030050 5 g Glass bottle
AC222030250 25 g Glass bottle
AC222031000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC222030050 Supplier Thermo Scientific Chemicals Supplier No. 222030050

CAS: 10147-37-2 | C3H7ClO2S | 142.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10147-37-2
Molecular Formula C3H7ClO2S
Molecular Weight (g/mol) 142.61
InChI Key DRINJBFRTLBHNF-UHFFFAOYSA-N
Synonym isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride
PubChem CID 82408
IUPAC Name propane-2-sulfonyl chloride
SMILES CC(C)S(=O)(=O)Cl

Specifications

Melting Point -47.0°C
Density 1.2730g/mL
Boiling Point 95.0°C (45.0 mmHg)
Flash Point 86°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHSO2Cl
Quantity 5 g
Beilstein 04,8
Specific Gravity 1.273
Formula Weight 142.61
Percent Purity 97%
Chemical Name or Material 2-Propanesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 233-415-2

RUO – Research Use Only