missing translation for 'onlineSavingsMsg'
Learn More

2-Propanesulfonyl chloride, 97%

Catalog No. AAL0243106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0243106 5 g
AAL0243114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0243106 Supplier Thermo Scientific Chemicals Supplier No. L0243106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10147-37-2 | C3H7ClO2S | 142.597 g/mol

2-Propanesulfonyl chloride is used as a pharmaceutical intermediate. Reductive cleavage of a secondary alcohol to methylene has been achieved by reduction of the 2-propanesulfonyl ester with lithium triethylborohydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Propanesulfonyl chloride is used as a pharmaceutical intermediate. Reductive cleavage of a secondary alcohol to methylene has been achieved by reduction of the 2-propanesulfonyl ester with lithium triethylborohydride.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. Store under dry inert gas. It is sensitive to moisture.

Chemical Identifiers

CAS 10147-37-2
Molecular Formula C3H7ClO2S
Molecular Weight (g/mol) 142.597
MDL Number MFCD00007453
InChI Key DRINJBFRTLBHNF-UHFFFAOYSA-N
Synonym isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride
PubChem CID 82408
IUPAC Name propane-2-sulfonyl chloride
SMILES CC(C)S(=O)(=O)Cl

Specifications

Melting Point -47°C
Density 1.273
Boiling Point 95°C (45 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.456
Quantity 5 g
UN Number UN3265
Beilstein 1747497
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 142.6
Percent Purity 97%
Chemical Name or Material 2-Propanesulfonyl chloride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 233-415-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only