missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Piperidone 98.0+%, TCI America™
Supplier: TCI America P0455500G
Specifications
| 2-Piperidone | |
| 256°C | |
| MFCD00006037 | |
| 2-piperidone, 2-piperidinone, delta-valerolactam, valerolactim, 5-pentanolactam, piperidinone, piperidon, piperidone, alpha-piperidone, piperidone-2 | |
| O=C1CCCCN1 | |
| 99.13 | |
| CHEBI:77761 | |
| ≥98.0% (GC) |
| 675-20-7 | |
| C5H9NO | |
| 500 g | |
| XUWHAWMETYGRKB-UHFFFAOYSA-N | |
| piperidin-2-one | |
| 12665 | |
| 99.13 | |
| Crystals |
Chemical Identifiers
| 675-20-7 | |
| 99.13 | |
| XUWHAWMETYGRKB-UHFFFAOYSA-N | |
| 12665 | |
| piperidin-2-one |
| C5H9NO | |
| MFCD00006037 | |
| 2-piperidone, 2-piperidinone, delta-valerolactam, valerolactim, 5-pentanolactam, piperidinone, piperidon, piperidone, alpha-piperidone, piperidone-2 | |
| CHEBI:77761 | |
| O=C1CCCCN1 |
Safety and Handling
RTECSNumber : TO0110000
TSCA : Yes