missing translation for 'onlineSavingsMsg'
Learn More

2-Piperidone 98.0+%, TCI America™

Catalog No. p-7134616
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P045525G 25 g
P0455100G 100 g
P0455500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P045525G Supplier TCI America Supplier No. P045525G
Only null left

Chemical Identifiers

CAS 675-20-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00006037
InChI Key XUWHAWMETYGRKB-UHFFFAOYSA-N
Synonym 2-piperidone, 2-piperidinone, delta-valerolactam, valerolactim, 5-pentanolactam, piperidinone, piperidon, piperidone, alpha-piperidone, piperidone-2
PubChem CID 12665
ChEBI CHEBI:77761
IUPAC Name piperidin-2-one
SMILES O=C1CCCCN1

Specifications

Boiling Point 256°C
Quantity 25 g
Formula Weight 99.13
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2-Piperidone
RTECSNumber TO0110000
TSCA Yes