missing translation for 'onlineSavingsMsg'
Learn More

2-piperidinoaniline, 97%, Thermo Scientific™

$22.56 - $22.56

Chemical Identifiers

CAS 39643-31-7
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00047467
InChI Key OYECAJPUPWFCSL-UHFFFAOYSA-N
Synonym 2-piperidinoaniline, 2-piperidin-1-yl aniline, 2-piperidin-1-yl-phenylamine, benzenamine, 2-1-piperidinyl, n-2-aminophenyl piperidine, 2-1-piperidinyl aniline, 2-1-piperidino aniline, 1-2-aminophenyl piperidine, 2-piperidylphenylamine, 2-piperdinoaniline
PubChem CID 458795
IUPAC Name 2-piperidin-1-ylaniline
SMILES NC1=CC=CC=C1N1CCCCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB02217DA
SDS View Documents
Thermo Scientific Chemicals
BTB02217DA
1 g
Each for $22.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

39643-31-7
176.26
OYECAJPUPWFCSL-UHFFFAOYSA-N
458795
NC1=CC=CC=C1N1CCCCC1
C11H16N2
MFCD00047467
2-piperidinoaniline, 2-piperidin-1-yl aniline, 2-piperidin-1-yl-phenylamine, benzenamine, 2-1-piperidinyl, n-2-aminophenyl piperidine, 2-1-piperidinyl aniline, 2-1-piperidino aniline, 1-2-aminophenyl piperidine, 2-piperidylphenylamine, 2-piperdinoaniline
2-piperidin-1-ylaniline

Specifications

39643-31-7
100.0
Amber Glass Bottle
MFCD00047467
2-piperidinoaniline, 2-piperidin-1-yl aniline, 2-piperidin-1-yl-phenylamine, benzenamine, 2-1-piperidinyl, n-2-aminophenyl piperidine, 2-1-piperidinyl aniline, 2-1-piperidino aniline, 1-2-aminophenyl piperidine, 2-piperidylphenylamine, 2-piperdinoaniline
NC1=CC=CC=C1N1CCCCC1
176.26
176.26
2-Piperidinoaniline
96.5
97%
C11H16N2
1 g
OYECAJPUPWFCSL-UHFFFAOYSA-N
2-piperidin-1-ylaniline
458795
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'