accessibility menu, dialog, popup

UserName

NH2N

CAS RN 39643-31-7

IUPAC Name: 2-(piperidin-1-yl)aniline
Synonyms: 2-(piperidin-1-yl)aniline
Molecular Weight (g/mol): 176.26
Molecular Formula: C11H16N2
InChi Key: OYECAJPUPWFCSL-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1N1CCCCC1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)