missing translation for 'onlineSavingsMsg'
Learn More

2-Piperidinemethanol 98.0+%, TCI America™

Catalog No. P101725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P101725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P101725G Supplier TCI America Supplier No. P101725G

Chemical Identifiers

CAS 3433-37-2
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00005987
InChI Key PRAYXGYYVXRDDW-UHFFFAOYSA-N
Synonym 2-piperidinemethanol, 2-hydroxymethylpiperidine, 2-hydroxymethyl piperidine, 2-piperidinylmethanol, piperidin-2-yl-methanol, piperidine-2-methanol, 2-piperidylmethan-1-ol, 2-piperidine methanol, pubchem6779
PubChem CID 94263
IUPAC Name piperidin-2-ylmethanol
SMILES C1CCNC(C1)CO

Specifications

Melting Point 67°C
Color White
Boiling Point 100°C
Quantity 25 g
UN Number 3263
Formula Weight 115.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Piperidinemethanol
EINECSNumber (5)-3729
TSCA No