accessibility menu, dialog, popup

UserName

HO(R)H2N

CAS RN 3433-37-2

IUPAC Name: (2R)-2-(hydroxymethyl)piperidin-1-ium
Synonyms: (2R)-2-(hydroxymethyl)piperidin-1-ium
Molecular Weight (g/mol): 116.18
Molecular Formula: C6H14NO
InChi Key: PRAYXGYYVXRDDW-ZCFIWIBFSA-O
SMILES: OC[C@H]1CCCC[NH2+]1
Boiling Point 
  • (1)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)