Learn More
2-Piperidinemethanol, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 168031000
| Quantity | 100g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 3433-37-2 | |
| 115.17 | |
| 2-piperidinemethanol, 2-hydroxymethylpiperidine, 2-hydroxymethyl piperidine, 2-piperidinylmethanol, piperidin-2-yl-methanol, piperidine-2-methanol, 2-piperidylmethan-1-ol, 2-piperidine methanol, pubchem6779 | |
| piperidin-2-ylmethanol |
| C6H13NO | |
| PRAYXGYYVXRDDW-UHFFFAOYSA-N | |
| 94263 | |
| C1CCNC(C1)CO |
Specifications
| 2-Piperidinemethanol | |
| 96.0 | |
| 100°C to 102°C (8.0 hPa) | |
| 96% min. (GC) | |
| C6H13NO | |
| 2-piperidinemethanol, 2-hydroxymethylpiperidine, 2-hydroxymethyl piperidine, 2-piperidinylmethanol, piperidin-2-yl-methanol, piperidine-2-methanol, 2-piperidylmethan-1-ol, 2-piperidine methanol, pubchem6779 | |
| C1CCNC(C1)CO | |
| 115.17 | |
| 115.17 |
| 3433-37-2 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| 100g | |
| PRAYXGYYVXRDDW-UHFFFAOYSA-N | |
| piperidin-2-ylmethanol | |
| 94263 | |
| 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning
EINECSNumber : 222-333-2
Promotions