missing translation for 'onlineSavingsMsg'
Learn More

2-Piperidinemethanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 168030250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC168030250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3433-37-2
115.17
2-piperidinemethanol, 2-hydroxymethylpiperidine, 2-hydroxymethyl piperidine, 2-piperidinylmethanol, piperidin-2-yl-methanol, piperidine-2-methanol, 2-piperidylmethan-1-ol, 2-piperidine methanol, pubchem6779
piperidin-2-ylmethanol
C6H13NO
PRAYXGYYVXRDDW-UHFFFAOYSA-N
94263
C1CCNC(C1)CO

Specifications

2-Piperidinemethanol
96.0
100°C to 102°C (8.0 hPa)
96% min. (GC)
C6H13NO
2-piperidinemethanol, 2-hydroxymethylpiperidine, 2-hydroxymethyl piperidine, 2-piperidinylmethanol, piperidin-2-yl-methanol, piperidine-2-methanol, 2-piperidylmethan-1-ol, 2-piperidine methanol, pubchem6779
C1CCNC(C1)CO
115.17
115.17
3433-37-2
100.0
Authentic
Glass bottle
25g
PRAYXGYYVXRDDW-UHFFFAOYSA-N
piperidin-2-ylmethanol
94263
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning

EINECSNumber : 222-333-2

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...