Learn More
2-Piperidinemethanol, 95%
CAS: 3433-37-2 | C6H13NO | 115.176 g/mol
$141.40 - $527.82
Chemical Identifiers
| CAS | 3433-37-2 |
|---|---|
| Molecular Formula | C6H13NO |
| Molecular Weight (g/mol) | 115.176 |
| MDL Number | MFCD00005987 |
| InChI Key | PRAYXGYYVXRDDW-UHFFFAOYSA-N |
| Synonym | 2-piperidinemethanol, 2-hydroxymethylpiperidine, 2-hydroxymethyl piperidine, 2-piperidinylmethanol, piperidin-2-yl-methanol, piperidine-2-methanol, 2-piperidylmethan-1-ol, 2-piperidine methanol, pubchem6779 |
| PubChem CID | 94263 |
| IUPAC Name | piperidin-2-ylmethanol |
| SMILES | C1CCNC(C1)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6017809
|
Thermo Scientific Chemicals
H6017809 |
10 g |
Each for $141.40
|
|
|||||
|
AAH6017818
|
Thermo Scientific Chemicals
H6017818 |
50 g |
Each for $527.82
|
|
|||||
Description
2-Piperidinemethanol was used in the synthesis of 3-phenyloctahydropyrido[2,1-c][1,4]oxazine hydrochloride and the 10R and 10S diastereomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Piperidinemethanol was used in the synthesis of 3-phenyloctahydropyrido[2,1-c][1,4]oxazine hydrochloride and the 10R and 10S diastereomers.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 3433-37-2 | |
| 115.176 | |
| PRAYXGYYVXRDDW-UHFFFAOYSA-N | |
| 94263 | |
| C1CCNC(C1)CO |
| C6H13NO | |
| MFCD00005987 | |
| 2-piperidinemethanol, 2-hydroxymethylpiperidine, 2-hydroxymethyl piperidine, 2-piperidinylmethanol, piperidin-2-yl-methanol, piperidine-2-methanol, 2-piperidylmethan-1-ol, 2-piperidine methanol, pubchem6779 | |
| piperidin-2-ylmethanol |
Specifications
| 3433-37-2 | |
| 100°C to 102°C | |
| MFCD00005987 | |
| 2-piperidinemethanol, 2-hydroxymethylpiperidine, 2-hydroxymethyl piperidine, 2-piperidinylmethanol, piperidin-2-yl-methanol, piperidine-2-methanol, 2-piperidylmethan-1-ol, 2-piperidine methanol, pubchem6779 | |
| PRAYXGYYVXRDDW-UHFFFAOYSA-N | |
| piperidin-2-ylmethanol | |
| 94263 | |
| 97% |
| 68°C to 70°C | |
| C6H13NO | |
| 10 g | |
| Slightly soluble in water. | |
| C1CCNC(C1)CO | |
| 115.176 | |
| 115.17 | |
| 2-Piperidinemethanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 222-333-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only