missing translation for 'onlineSavingsMsg'
Learn More

2-Piperidinemethanol, 95%

Catalog No. AAH6017809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6017809 10 g
AAH6017818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6017809 Supplier Thermo Scientific Chemicals Supplier No. H6017809

CAS: 3433-37-2 | C6H13NO | 115.176 g/mol

2-Piperidinemethanol was used in the synthesis of 3-phenyloctahydropyrido[2,1-c][1,4]oxazine hydrochloride and the 10R and 10S diastereomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Piperidinemethanol was used in the synthesis of 3-phenyloctahydropyrido[2,1-c][1,4]oxazine hydrochloride and the 10R and 10S diastereomers.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3433-37-2
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00005987
InChI Key PRAYXGYYVXRDDW-UHFFFAOYSA-N
Synonym 2-piperidinemethanol, 2-hydroxymethylpiperidine, 2-hydroxymethyl piperidine, 2-piperidinylmethanol, piperidin-2-yl-methanol, piperidine-2-methanol, 2-piperidylmethan-1-ol, 2-piperidine methanol, pubchem6779
PubChem CID 94263
IUPAC Name piperidin-2-ylmethanol
SMILES C1CCNC(C1)CO

Specifications

Melting Point 68°C to 70°C
Boiling Point 100°C to 102°C
Quantity 10 g
Solubility Information Slightly soluble in water.
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material 2-Piperidinemethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-333-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only