missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Piperazinone 98.0+%, TCI America™
$42.87 - $124.89
Chemical Identifiers
| CAS | 5625-67-2 |
|---|---|
| Molecular Formula | C4H9N2O |
| Molecular Weight (g/mol) | 101.13 |
| MDL Number | MFCD01318687 |
| InChI Key | IWELDVXSEVIIGI-UHFFFAOYSA-O |
| Synonym | 2-piperazinone, 2-oxopiperazine, piperazine-2-one, piperazinone, 2-piperadone, oxopiperazine, piperazone, keto piperazine, 3-oxopiperazine, 2-ketopiperazine |
| PubChem CID | 231360 |
| IUPAC Name | 3-oxopiperazin-1-ium |
| SMILES | O=C1C[NH2+]CCN1 |
Chemical Identifiers
| 5625-67-2 | |
| 101.13 | |
| IWELDVXSEVIIGI-UHFFFAOYSA-O | |
| 231360 | |
| O=C1C[NH2+]CCN1 |
| C4H9N2O | |
| MFCD01318687 | |
| 2-piperazinone, 2-oxopiperazine, piperazine-2-one, piperazinone, 2-piperadone, oxopiperazine, piperazone, keto piperazine, 3-oxopiperazine, 2-ketopiperazine | |
| 3-oxopiperazin-1-ium |
Specifications
| 5625-67-2 | |
| 164°C | |
| MFCD01318687 | |
| 2-piperazinone, 2-oxopiperazine, piperazine-2-one, piperazinone, 2-piperadone, oxopiperazine, piperazone, keto piperazine, 3-oxopiperazine, 2-ketopiperazine | |
| O=C1C[NH2+]CCN1 | |
| 101.13 | |
| 100.12 | |
| Crystalline Powder |
| 134°C | |
| C4H9N2O | |
| 1 g | |
| IWELDVXSEVIIGI-UHFFFAOYSA-O | |
| 3-oxopiperazin-1-ium | |
| 231360 | |
| ≥98.0% (GC) | |
| 2-Piperazinone |
Safety and Handling
TSCA : No