accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 5625-67-2

ONH2HN

CAS RN 5625-67-2

IUPAC Name: 3-oxopiperazin-1-ium
Synonyms: 3-oxopiperazin-1-ium
Molecular Weight (g/mol): 101.13
Molecular Formula: C4H9N2O
InChi Key: IWELDVXSEVIIGI-UHFFFAOYSA-O
SMILES: O=C1C[NH2+]CCN1
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.